HPLC Analysis of N-Acetylphenylalanine Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of N-Acetylphenylalanine Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 23 C
mobile phase [A] 0.1% triethylamine acetate, pH 3.8; [B] ethanol; (20:80, A:B)
flow rate 1 ml/min
sample 5 mg/mL in methanol
injection 5 µl
detector UV, 225 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, All Amino Acids, Native Amino Acids
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC