HPLC Analysis of N-Desmethylnefopam Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of N-Desmethylnefopam Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 µm particles (15024AST)
column temp. 25 °C
mobile phase [A] methanol; [B] acetic acid; [C] ammonium hydroxide; (100:0.1:0.02, A:B:C)
flow rate 1.2 mL/min
sample 2 mg/mL in methanol
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Analgesics, Pharmaceutical (small molecule), All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC