Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Nonionic Surfactants, 9-10 Ethoxy Groups (Octylphenol) on SUPELCOSIL™ LC-Diol
HPLC Analysis of Nonionic Surfactants, 9-10 Ethoxy Groups (Octylphenol) on SUPELCOSIL™ LC-Diol
Conditions
column SUPELCOSIL LC-Diol, 25 cm x 4.6 mm I.D., 5 µm particles (58201)
column temp. 35 C
mobile phase [A] hexane:methylene chloride (95:5); [B] hexane:methylene chloride:methanol (50:40:10)
flow rate 1 ml/min
injection 10 muLmethylene chloride containing 20 mug surfactant
detector UV, 280 nm
Description
Categories Analytical Chromatography, Surfactants
Featured Industry Chemicals and Industrial Polymers
Legal Information SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC