HPLC Analysis of Norgestrel Enantiomers on Astec® CYCLOBOND® II
HPLC Analysis of Norgestrel Enantiomers on Astec® CYCLOBOND® II
Conditions
column CYCLOBOND II, 25 cm x 4.6 mm I.D., 5 µm particles (41020AST)
column temp. 22 °C
mobile phase [A] water; [B] acetonitrile; (70:30, A:B)
flow rate 0.8 mL/min
sample norgestrel, 1 mg/mL in methanol
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Clinical, Chiral Applications, Hormones, hormones, Clinical, Forensic & Toxicology
Featured Industry Clinical
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC