HPLC Analysis of NSAIDs on Discovery® Zr-Carbon C18
HPLC Analysis of NSAIDs on Discovery® Zr-Carbon C18
Conditions
column Discovery Zr-Carbon C18, 15 cm x 4.6 mm I.D., 3 µm particles (65706-U)
column temp. 80 °C
mobile phase [A] 50 mM phosphoric acid (pH 1.75): [B] acetonitrile; (50:50, A:B)
flow rate 4 mL/min
sample ketoprofen, ibuprofen, naproxen, each 1 mg/mL
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anti-inflammatories
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC