HPLC Analysis of Nucleosides on Ascentis® Express HILIC
HPLC Analysis of Nucleosides on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 µm particles (53939-U)
column temp. 35 °C
mobile phase [A] 100 mM ammonium formate, pH 3.0 with concentrated formic acid: [B] acetonitrile; (10:90, A:B)
flow rate 0.4 mL/min
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Nucleic Acids