HPLC Analysis of Nucleosides on TSKgel® Boronate-5PW, Isocratic Mode
HPLC Analysis of Nucleosides on TSKgel® Boronate-5PW, Isocratic Mode
Conditions
column TSKgel Boronate-5PW, 7.5 cm x 7.5 mm I.D., 10 µm particles (813066)
mobile phase 0.1 M phosphate buffer, pH 8.0
flow rate 1 ml/min
detector UV, 280 nm
Description
Categories Analytical Chromatography, Nucleosides
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.