HPLC Analysis of of Fab and Fc Fragments on TSKgel® Butyl-NPR
HPLC Analysis of of Fab and Fc Fragments on TSKgel® Butyl-NPR
Conditions
column TSKgel Butyl-NPR, 3.5 cm x 4.6 mm I.D., 2.5 µm particles (814947)
column temp. 30 °C
mobile phase [A] 2 M ammonium sulfate, 20 mM Tris, pH 7; [B] 20 mM Tris, pH 7
gradient 10 to 100% B in 34 min
flow rate 1 mL/min
Description
Categories Analytical Chromatography, Antibodies
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.