HPLC Analysis of Oligonucleotides on Proteomix® SAX
HPLC Analysis of Oligonucleotides on Proteomix® SAX
Conditions
column Proteomix SAX-NP5, 25 cm x 4.6 mm I.D., 5 µm particles (Z777229)
column temp. 25 °C
mobile phase A: 20 mM Tris, pH 8.0 B: A + 0.5 M NaCl
gradient 0 - 100% B in 30 minutes
flow rate 0.5 mL/min
pressure 1073 psi (74 bar)
sample (10 mM each oligonucleotide in water)
injection 5 µL
detector UV 260 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Proteomix is a registered trademark of Sepax Technologies