HPLC Analysis of Organic Acids Commonly Found in Wine and Beer on TSKgel® OApak-P and OApak-A
HPLC Analysis of Organic Acids Commonly Found in Wine and Beer on TSKgel® OApak-P and OApak-A
Conditions
column TSKgel OApak-P Guard Column, 4 cm x 6 mm I.D., 10 µm particles (816654)
column TSKgel OApak-A, 30 cm x 7.8 mm I.D., 5 µm particles (816653)
column temp. 60 °C
mobile phase 0.75 mM sulfuric acid
flow rate 0.8 mL/min
sample organic acid mixture, see figure for concentrations
injection 20 µL
detector conductivity
Description
Categories Analytical Chromatography, Organic Acids
Featured Industry Food and Beverages
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.