HPLC Analysis of Organic Acids on Discovery® C18
HPLC Analysis of Organic Acids on Discovery® C18
Conditions
column Discovery C18, 5 cm x 4.6 mm I.D., 5 µm particles (504947)
column temp. 20 °C
mobile phase [A] 0.1% TFA in [B] water:methanol; (60:40, A:B)
flow rate 2.0 mL/min
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Organic Acids
Featured Industry Chemicals and Industrial Polymers
Food and Beverages
Vitamins, Nutraceuticals, and Natural Products
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC