HPLC Analysis of Organic Acids on Ascentis® Express RP-Amide
HPLC Analysis of Organic Acids on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.7 µm particles (53913-U)
column temp. 35 °C
mobile phase [A] 0.1% (v/v) TFA in [B] water
flow rate 0.3 mL/min
sample 0.2 - 4 g/L in mobile phase
injection 1 µL
detector UV, 210 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express RP-Amide for the analysis of organic acids under 100% aqueous conditions.
Categories Analytical Chromatography, Organic Acids
Featured Industry Chemicals and Industrial Polymers
Food and Beverages
Vitamins, Nutraceuticals, and Natural Products