HPLC Analysis of Organo-acid Pesticides on Discovery® C18
HPLC Analysis of Organo-acid Pesticides on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 20 °C
mobile phase [A] 25 mM potassium phosphate, (pH 2.3): [B] acetonitrile; (60:40, A:B)
flow rate 2 mL/min
injection 1 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Environmental Analysis, Food & Beverage Analysis, Pesticides & Herbicides, Organics, Pesticide and Metabolite Residues, Chromatograms, Applications (Chromatograms), HPLC Chromatograms
Featured Industry Environmental
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC