HPLC Analysis of Organophosphorus Pesticides on Discovery® RP-Amide C16
HPLC Analysis of Organophosphorus Pesticides on Discovery® RP-Amide C16
Conditions
column Discovery RP-AmideC16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. 20 °C
mobile phase [A] acetonitrile: [B] water; (60:40, A:B)
flow rate 1 mL/min
injection 1 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Environmental Analysis, Pesticides & Herbicides, Organics, Chromatograms, HPLC Chromatograms
Featured Industry Environmental
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC