Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Ovalbumin and BSA Charge Variants on Proteomix® SAX-NP3
HPLC Analysis of Ovalbumin and BSA Charge Variants on Proteomix® SAX-NP3
Conditions
column Proteomix SAX-NP3, 5 cm x 4.6 mm I.D., 3 µm particles (Z777219)
mobile phase 20 mM Tris buffer, pH 8.0
gradient 0 - 0.3 M NaCl in 15 min
flow rate 0.5 ml/min
pressure 1600 psi (110 bar)
detector UV, 214 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Proteomix is a registered trademark of Sepax Technologies