HPLC Analysis of Oxazepam Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Oxazepam Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 25 °C
mobile phase methanol
flow rate 1 mL/min
injection 1 µL
sample gas 1 mg/mL in methanol
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Anticonvulsant/antiepiletic/antiseizure, All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC