HPLC Analysis of Oxazepam Enantiomers on Astec® CYCLOBOND® I 2000 DNP
HPLC Analysis of Oxazepam Enantiomers on Astec® CYCLOBOND® I 2000 DNP
Conditions
column CYCLOBOND I 2000 DNP, 25 cm x 4.6 mm I.D., 5 µm particles (25024AST)
column temp. 5 °C
mobile phase [A] acetonitrile; [B] 20 mM ammonium phosphate, pH 2.9; (20:80, A:B)
flow rate 1 mL/min
sample oxazepam, 1 mg/mL in acetonitrile:water (50:50)
injection 3 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Anxiolytics, Sedatives
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC