HPLC Analysis of Oxprenolol Enantiomers on Astec® Cellulose DMP
HPLC Analysis of Oxprenolol Enantiomers on Astec® Cellulose DMP
Conditions
column Astec Cellulose DMP, 25 cm x 4.6 mm I.D., 5 µm particles (51099AST)
column temp. 30 °C
mobile phase [A] heptane; [B] isopropanol; [C] diethylamine; (80:20:0.1, A:B:C)
flow rate 1 mL/min
sample oxprenolol, 1 g/L in 50:50 heptane:isopropanol
injection 15 µL
detector UV, 254 nm
Description
Analysis Note Oxprenolol enantiomers are resolved by normal phase chromatography.
Categories Analytical Chromatography, Antihypertensives
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Oxprenolol is a nonselective beta-blocker and is used to treat angina and hypertension.