HPLC Analysis of p-Fluorophenyl Methyl Sulfoxide Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of p-Fluorophenyl Methyl Sulfoxide Enantiomers on Astec® CHIROBIOTIC® TAG
Conditions
column CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 µm particles (14024AST)
mobile phase [A] hexane; [B] ethanol; (50:50, A:B)
flow rate 2 ml/min
detector UV, polarimetry to confirm elution order
Description
Analysis Note Details: A. Berthod et al, J. Chromatogr A, 955 (2002) 53-69.
Categories Analytical Chromatography, Chiral Applications, Organic Chemicals, Fluorinated compounds, Chemicals and Industrial Polymers, Other Organic Compound Applications
Featured Industry Chemicals and Industrial Polymers
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC