Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of p-Toluenesulfonic Acid (PTSA) and Pyrimethamine on SeQuant ZIC-HILIC
HPLC Analysis of p-Toluenesulfonic Acid (PTSA) and Pyrimethamine on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 µm, 200 Ã… (1.50455.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] ammonium acetate; (90:10, A:B)
flow rate 0.5 ml/min
injection 1 µl
detector 254 nm
Description
Featured Industry Chemicals and Industrial Polymers