Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of PAHs in Water on SUPELCOSIL™ LC-PAH after SPME using 100 μm PDMS Fiber
HPLC Analysis of PAHs in Water on SUPELCOSIL™ LC-PAH after SPME using 100 μm PDMS Fiber
Conditions
sample/matrix 5 µl PAH mix (Cat. No. 4-8743) in 5 mL water
desorption process static, 200 µl acetonitrile:water, 40:60, 2 min
extraction immersion, 30 min (rapid stirring)
SPME fiber polydimethylsiloxane, 100 µm (57301)
column SUPELCOSIL LC-PAH, 15 cm x 4.6 mm I.D., 5 µm particles (58318)
mobile phase [A] acetonitrile:[B] water
gradient 0 min - 50% A; 5 min - 50% A; 30 min - 100% A; 45 min - 100% A
flow rate 0-2 min: 0.2 ml/min 2-45 min: 1.0 ml/min
detector UV, 254 nm
Description
Categories Analytical Chromatography, Food & Beverage Analysis, Environmental Analysis, SPME Applications, Environmental, Processing/Packaging Contaminants, Organics, Semivolatiles, PAHs, Chromatograms, Applications (Chromatograms), HPLC Chromatograms
Featured Industry Environmental
Legal Information SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC
suitability application for SPME, application for HPLC