HPLC Analysis of Pain Relievers on Ascentis® Express C18
HPLC Analysis of Pain Relievers on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm x 4.6 mm I.D., 5 µm particles (50536-U)
column temp. 45 °C
mobile phase [A] water with 3% acetic acid; [B] methanol with 3% acetic acid; (79:21, A:B)
flow rate 2.0 mL/min
pressure 3290 psi (227 bar)
sample 100 - 360 µg/mLin mobile phase
injection 10 µL
detector UV,275 nm
Description
Analysis Note This application shows the improvements in efficiency that may be obtained for USP methods using modern Fused-Core technology.
Categories Analytical Chromatography, Clinical, analgesics, Analgesics
Featured Industry Clinical
Pharmaceutical (small molecule)