HPLC Analysis of Paraben Preservatives on Discovery® C18
HPLC Analysis of Paraben Preservatives on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 20 °C
mobile phase [A] acetonitrile: [B] water, (30:70, A:B)
flow rate 1 mL/min
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Preservatives
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC