HPLC Analysis of Paraben Preservatives on Discovery® Cyano
HPLC Analysis of Paraben Preservatives on Discovery® Cyano
Conditions
column Discovery Cyano, 15 cm x 4.6 mm I.D., 5 µm particles (59356-U)
column temp. 20 °C
mobile phase [A] acetonitrile; [B] water; (40:60, A:B)
flow rate 1 mL/min
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Preservatives
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC