HPLC Analysis of pBr322 DNA-Hae III Digest on TSKgel® DEAE-NPR
HPLC Analysis of pBr322 DNA-Hae III Digest on TSKgel® DEAE-NPR
Conditions
column TSKgel DEAE-NPR, 3.5 cm x 4.6 mm I.D., 2.5 mum particles (with 5 mm DEAE-NPR guard column) (813075)
column temp. ambient
mobile phase [A] 1 M sodium chloride/25 mm Tris-HCl, pH 9.0; [B] 25 mM Tris-HCl, pH 9.0
flow rate 1 mL/min
injection 20 µL of 50 µg/mL Hae III digest of pBR322 DNA
detector UV, 260 nm
Description
Categories Analytical Chromatography, Nucleic Acids
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation