HPLC Analysis of Pegylated Exenatide on Zenix® SEC-300 and Zenix®-C SEC-300
HPLC Analysis of Pegylated Exenatide on Zenix® SEC-300 and Zenix®-C SEC-300
Conditions
column Zenix-C SEC-300, 30 cm x 7.8 mm, I.D., 3 µm particles (Z777092)
column Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 µm particles (Z777033)
column temp. 25 °C
mobile phase (A) 50 mM Ammonium acetate: (B) acetonitrile; (90:10, A:B, v/v)
flow rate 0.5 mL/min
pressure 609 psi (42 bar)
sample 3.3 mg/ml PEG-Exanatide in water (PEG 23 kD)
injection 15 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
Legal Information Zenix is a registered trademark of Sepax Technologies