HPLC Analysis of Penicillin G and Phenoxymethylpenicillinic Acid on Ascentis® C18
HPLC Analysis of Penicillin G and Phenoxymethylpenicillinic Acid on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium acetate (pH 4.5 acetic acid): [B] acetonitrile, (75:25, A:B)
flow rate 1 mL/min
sample 100 µg/mL each in mobile phase
injection 10 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Clinical, Pharmaceutical Analysis, antibiotics/antimicrobials, Antibiotics, Antibiotics, Antimicrobials
Featured Industry Clinical