HPLC Analysis of Pentylbenzene and o-Terphenyl on Ascentis® Express Biphenyl
HPLC Analysis of Pentylbenzene and o-Terphenyl on Ascentis® Express Biphenyl
Conditions
column Ascentis Express Biphenyl, 10cm x 2.1mm, 2.7 µm particles (64065-U)
mobile phase [A]:water [B]: methanol; (20:80 A:B)
flow rate 0.5 ml/min
pressure 5400 psi (372 bar)
sample Toluene(500 µg/mL) n-pentylbenzene (500 µg/mL), o-Terphenylene (50 µg/mL).    Prepared in 50:50 A:B
injection 1 µl
detector UV, 254 nm
Description
Analysis Note This application demonstrates the separation of pentylbenzene and o-Terphenyl using Ascentis Express Biphenyl. It demonstrates the applicability of this phase to seperating substituted aromatic compounds, and also its potential utility in positional isomer seperations of Terphenyl. This set of analytes is used as a probe of pi acidity of a stationary phase.
Categories Analytical Chromatography, Pharmaceuticals, Aromatics
Featured Industry Chemicals and Industrial Polymers
Pharmaceutical (small molecule)