HPLC Analysis of Pepides, Alberta Mix, on Discovery® BIO Wide Pore C18, No TFA
HPLC Analysis of Pepides, Alberta Mix, on Discovery® BIO Wide Pore C18, No TFA
Conditions
column Discovery BIO Wide Pore C18, 15 cm x 2.1 mm I.D., 3 µm particles (567202-U)
column temp. ambient
mobile phase [A] 25 mM formic acid in water, [B] (25 mM formic acid in water):(20 mM formic acid in acetonitrile, (50:50)
flow rate 0.20 mL/min
injection 1 µL
detector +ES
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC