Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Pepides, Alberta Mix on Discovery® BIO Wide Pore C5
HPLC Analysis of Pepides, Alberta Mix on Discovery® BIO Wide Pore C5
Conditions
column Discovery BIO Wide Pore C5, 15cm x 2.1mm I.D., 5 µm particles (568402-U)
column temp. 35 C
mobile phase [A] 25mM formic acid in water; [B] (50:50) 25mM formic acid in water, 20mM formic acid in acetonitrile
flow rate 0.20 ml/min
sample RP Peptide Ionic Interactions Standard, p/n RPS-I0020 (Alberta Peptide Institute)
injection 0.5 µl (~0.25 mug each peptide)
detector UV, 215 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC