HPLC Analysis of Peptide Antibiotics on Discovery® C8
HPLC Analysis of Peptide Antibiotics on Discovery® C8
Conditions
column Discovery C8, 15 cm x 4.6 mm I.D., 5 µm particles (59353)
column temp. 35 °C
mobile phase [A] 25mM potassium phosphate, pH 3; [B] acetonitrile, (60:40, A:B)
flow rate 1 mL/min
injection 10 µL10 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antibiotics
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC