Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Peptide Mix, Complex, on Ascentis® C18
HPLC Analysis of Peptide Mix, Complex, on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 2.1 mm I.D., 5 µm particles (581304-U)
column temp. 35 C
mobile phase [A] 25 mM formic acid in water; [B]: 50:50 (v/v) 25 mM formic acid in water:25 mM formic acid in acetonitrile
gradient 10 to 64% B in 90 min
flow rate 0.2 ml/min
sample tryptic digest of carboxymethylated protein mixture 2.4 g/L in 0.1% formic acid
injection 10 µl
detector ESI (+), full scan
Description
Categories Analytical Chromatography, Peptides