HPLC Analysis of Peptides on Discovery® BIO Wide Pore C18, Preparative Scale
HPLC Analysis of Peptides on Discovery® BIO Wide Pore C18, Preparative Scale
Conditions
column Discovery BIO Wide Pore C18, 15 cm x 10 mm I.D., 10 µm particles (567208-U)
column temp. 30 °C
mobile phase [A] (80:20), 0.1% TFA in water:0.1% TFA in acetonitrile; [B] (66:34), 0.1% TFA in water:0.1% TFA in acetonitrile
flow rate 4.7 mL/min
sample Cat. No. P2693
injection 24.5 µL
detector UV, 215 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC