HPLC Analysis of Peptides on Discovery® BIO Wide Pore C18, 28-38 Min. Window
HPLC Analysis of Peptides on Discovery® BIO Wide Pore C18, 28-38 Min. Window
Conditions
column Discovery BIO Wide Pore C18, 15cm x 4.6mm I.D., 5 µm particles (568222-U)
column temp. 30 °C
mobile phase [A] (95:5), 0.1% TFA in water:0.1% TFA in acetonitrile; [B] (50:50), 0.1% TFA in water:0.1% TFA in acetonitrile
gradient 0-100% B in 65 min
flow rate 1 mL/min
injection 50µL carboxymethylated apohemoglobin trypic digest in 50 mM ammonium bicarbonate
detector UV, 215 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC