Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Peptides on Proteomix® SCX-NP3, Volatile Buffer, ELSD and UV Detection
HPLC Analysis of Peptides on Proteomix® SCX-NP3, Volatile Buffer, ELSD and UV Detection
Conditions
column Proteomix SCX-NP3, 5 cm x 4.6 mm I.D., 3 µm particles (Z777151)
mobile phase (A) 5 mM ammonium acetate; (B) 50 mM ammonium acetate:acetonitrile (4:1, v:v)
gradient 0 to 50% B in 20 min
flow rate 0.6 ml/min
sample 0.1 mg/mL
injection 5 µl
detector ELSD (SofTA) and UV, 280 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Proteomix is a registered trademark of Sepax Technologies