Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Pesticides on Ascentis® C8 (15 cm x 4.6 mm x 5 μm)
HPLC Analysis of Pesticides on Ascentis® C8 (15 cm x 4.6 mm x 5 μm)
Conditions
column Ascentis C8, 15 cm x 4.6 mm I.D., 5 µm particles (581424-U)
column temp. 35 C
mobile phase [A] water; [B] acetonitrile
gradient 35 to 75% B in 13 min; held for 5 min
flow rate 1.0 ml/min
sample as indicated in 2.2% 90:10 acetonitrile:acetone in DI water
injection 100 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Food & Beverage Analysis, Environmental Analysis, Organics, Pesticides & Herbicides, Pesticide and Metabolite Residues, Applications (Chromatograms), Chromatograms, HPLC Chromatograms
Featured Industry Environmental