HPLC Analysis of Phenolic Antioxidants on Ascentis® C8
HPLC Analysis of Phenolic Antioxidants on Ascentis® C8
Conditions
column Ascentis C8, 15 cm x 4.6 mm I.D., 5 µm particles (581424-U)
column temp. 30 C
mobile phase [A] 5% acetic acid; [B] (50:50) methanol:acetonitrile
gradient 30 to 100% B in 10 min; held for 5 min
flow rate 2.0 ml/min
sample as indicated in 50:50 acetonitrile:2-propanol
injection 10 µl
detector UV, 280 nm
Description
Categories Analytical Chromatography, Antioxidants
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Vitamins, Nutraceuticals, and Natural Products