Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Phenolic Antioxidants on Ascentis® Express C18 5 μm
HPLC Analysis of Phenolic Antioxidants on Ascentis® Express C18 5 μm
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 5 µm particles (50537-U)
column temp. 40 C
mobile phase [A] water; [B] acetonitrile; [C] 2-propanol;
gradient Constant 43% C; 37 to 55% B in 10 min; held at 55% B for 5 min
flow rate 1.2 mL/min
pressure 1800 psi (124 bar)
sample 5 standards at 20 µg/mL
injection 20 µl
detector UV, 280 nm
Description
Analysis Note Phenolic Antioxidants are added to polyers for controlling their degradation process. Irgafos 168, Irganox 1010, Ethanox 330, Irganox 1076, BHT, and Irgafos 168 are often used as antioxidants to prevent the degradation of polypropylene polymer forµlations. This HPLC method is for the quantitation of an additive or mix of additives in a polymeric material.
Featured Industry Chemicals and Industrial Polymers