HPLC Analysis of Phenols on SUPELCOSIL™ LC-8
HPLC Analysis of Phenols on SUPELCOSIL™ LC-8
Conditions
column SUPELCOSIL LC-8, 15 cm x 4.6 mm I.D., 5 µm particles (58220-U)
column temp. 30 C
mobile phase [A] 1% acetic acid/methanol: [B] 1% acetic acid/water
gradient 35:65 to 100:0 (20 min), return to 35:65 (5 min)
flow rate 1.5 ml/min
detector UV, 280 nm
Description
Categories Analytical Chromatography, Phenols
Featured Industry Chemicals and Industrial Polymers
Legal Information SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC