HPLC Analysis of Phensuximide Enantiomers on Astec® CYCLOBOND® I 2000 DMP
HPLC Analysis of Phensuximide Enantiomers on Astec® CYCLOBOND® I 2000 DMP
Conditions
column CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 µm particles (20724AST)
column temp. 22 °C
mobile phase [A] isopropanol; [B] hexane; (80:20, A:B)
flow rate 1 mL/min
sample 4 mg/mL in methanol
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Anticonvulsant/antiepiletic/antiseizure, All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC