Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Phenylsuccinic Anhydride Enantiomers on Astec® (R,R) P-CAP™-DP
HPLC Analysis of Phenylsuccinic Anhydride Enantiomers on Astec® (R,R) P-CAP™-DP
Conditions
column Astec (R,R) P-CAP-DP, 25 cm x 4.6 mm I.D., 5 µm particles (35024AST)
column temp. 25 C
mobile phase [A] acetonitrile; [B] methanol; [C] trifluoroacetic acid; (99:1/0.1 (A:B)
flow rate 1 ml/min
sample 1 mg/mL in methanol
injection 1 µl
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Organic Chemicals, Anhydrides, Chemicals and Industrial Polymers, Other Organic Compound Applications
Featured Industry Chemicals and Industrial Polymers
Legal Information P-CAP is a trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC