Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Pilocarpine, Betaxolol, and Benzalkonium Chloride on Ascentis® Express HILIC
HPLC Analysis of Pilocarpine, Betaxolol, and Benzalkonium Chloride on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 15 cm x 4.6 mm I.D., 2.7 µm particles (53981-U)
column temp. 35 C
mobile phase [A] 20 mM ammonium acetate; [B] acetonitrile; (10:90, A:B)
flow rate 1 ml/min
sample 10 mg/L in acetonitrile
injection 2 µl
detector ESI (+), XIC m/z 304, 332 (BAC), 209 (pilocarpine), 308 (betaxolol)
Description
Categories Analytical Chromatography, Alkaloids, Antihypertensives, Surfactants