HPLC Analysis of Pipecolic Acid Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Pipecolic Acid Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 25 °C
mobile phase [A] water; [B] acetonitrile; (50:50, A:B)
flow rate 1 mL/min
sample 5 mg/mL in methanol
injection 1 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Chiral Applications, Clinical, Clinical, Forensic & Toxicology, Carboxylic acids, carboxylic acids, Chemicals and Industrial Polymers, Biomarkers
Featured Industry Clinical
Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC