HPLC Analysis of Plasmids on TSKgel® DNA-NPR
HPLC Analysis of Plasmids on TSKgel® DNA-NPR
Conditions
column TSKgel DNA-NPR, 7.5 cm x 4.6 mm, 2.5 µm particles (818249)
mobile phase [A] 0.02 M Tris, pH 9.0; [B] 0.02 M Tris, 1.0 M NaCl, pH 9.0
gradient 50 to 60% B in 10 column volumes
flow rate 1 mL/min
sample PUC19 plasmid
detector UV, 260 nm
Description
Categories Analytical Chromatography, Nucleic Acids
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.