HPLC Analysis of Polar Compounds on Ascentis® Express RP-Amide Particles
HPLC Analysis of Polar Compounds on Ascentis® Express RP-Amide Particles
Conditions
column Ascentis Express RP-Amide, 10 cm x 4.6 mm I.D., 5 µm particles (50773-U)
column temp. 26 °C
mobile phase [A] 20 mM potassium phosphate, pH 7 [B] acetonitrile (70:30, A:B)
flow rate 4.0 mL/min
sample 0.04-0.32 mg/mL in 50/50 water/acetonitrile
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Amides, Amines