Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Polar Neutral and Acidic Amino Acids on Ascentis® Si (Silica)
HPLC Analysis of Polar Neutral and Acidic Amino Acids on Ascentis® Si (Silica)
Conditions
column Ascentis Si, 10 cm x 2.1 mm I.D., 5 µm particles (581508-U)
column temp. 35 C
mobile phase [A] 100 mM ammonium formate, pH 3.0, with formic acid; [B] water, [C] acetonitrile
gradient 5% A constant; 10% B, 85% C to 16% B, 79% C in 20 min
flow rate 0.3 ml/min
sample 100 µg/mL in 10:90, (50 mM ammonium formate / formic acid, pH 3.0):acetonitrile
injection 2 µl
detector ESI (+), full scan
Description
Categories Analytical Chromatography, Amino Acids