HPLC Analysis of Polyethylene Glycol 200 on TSKgel® G-OLIGO-PW
HPLC Analysis of Polyethylene Glycol 200 on TSKgel® G-OLIGO-PW
Conditions
column TSKgel G-OLIGO-PW, 30 cm x 7.8 mm I.D., 6 µm particles (808031)
mobile phase distilled water
flow rate 1 ml/min
detector RI
Description
Categories Analytical Chromatography, Glycols, Diols
Featured Industry Chemicals and Industrial Polymers
Legal Information TSKgel is a registered trademark of Tosoh Corporation