Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)
HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)
Conditions
sample/matrix Nero d'Avola wine sample filtered prior injection through a 0.45 µm nylon membrane
column Discovery HS C18, 15 cm x 2.1 mm I.D., 3 µm particles (569255-U)
column temp. 30 C
mobile phase [A] water, pH 3 with formic acid; [B] acetonitrile, pH 3 with formic acid
flow rate 0.2 ml/min
injection 5 µl
detector UV, 280 nm
Description
Categories Analytical Chromatography, Clinical, Food & Beverage Analysis, phenols, Flavors, Fragrances, Beverage Testing, flavors/fragrances, Beer, Wine, and Spirits, Wine
Featured Industry Food and Beverages
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC