HPLC Analysis of Proglumide Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Proglumide Enantiomers on Astec® CHIROBIOTIC® V
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. 25 °C
mobile phase [A] 13 mM ammonium acetate; [B] acetonitrile; (90:10, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 1 µL
detector UV, 225 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antiulcer Compounds, Pharmaceutical (small molecule), Antiulcer Drugs, All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC