HPLC Analysis of Proline Enantiomers on Astec® CHIROBIOTIC® T, Fast Analysis
HPLC Analysis of Proline Enantiomers on Astec® CHIROBIOTIC® T, Fast Analysis
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 25 °C
mobile phase water
flow rate 0.65 mL/min
sample 1 mg/mL in methanol
injection 10 µL
detector UV, 210 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, All Amino Acids, Native Amino Acids
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC